missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GNE Mouse anti-Human, Unconjugated, Clone: 2A11E10, Proteintech
Mouse Monoclonal Antibody
1710.33 NOK - 4836.10 NOK
Specifications
| Antigen | GNE |
|---|---|
| Clone | 2A11E10 |
| Concentration | 1000 μg/mL |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
16860396
|
Proteintech
67649-1-IG-20UL |
20 μL |
1710.33 NOK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
16850396
|
Proteintech
67649-1-IG-150UL |
150 μL |
4836.10 NOK
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
The GNE antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GNE. This antibody recognizes human antigen. The GNE antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
| GNE | |
| 1000 μg/mL | |
| Monoclonal | |
| Mouse | |
| 10020 | |
| Primary |
| 2A11E10 | |
| Western Blot | |
| Unconjugated | |
| Human | |
| IgG1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title