Learn More
GN44028, MedChemExpress
Product Code. 16417097
Encompass_Preferred
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
Product Code. 16417097
Brand: Medchem Express HY110266
MedChemExpress GN44028 is a potent and orally active hypoxia inducible factor (HIF)-1α inhibitor, with an IC50 of 14 nM. GN44028 inhibits hypoxia-induced HIF-1α transcriptional activity without suppressing HIF-1α mRNA expression, HIF-1α protein accumulation, or HIF-1α/HIF-1β heterodimerization. GN44028 can be used in the research of cancers.
GN44028 is a potent and orally active hypoxia inducible factor (HIF)-1α inhibitor, with an IC50 of 14 nM. GN44028 inhibits hypoxia-induced HIF-1α transcriptional activity without suppressing HIF-1α mRNA expression, HIF-1α protein accumulation, or HIF-1α/HIF-1β heterodimerization. GN44028 can be used in the research of cancers.
Specifications
Chemical Name or Material GN44028
CAS 1421448-26-1
Color Off-White
Purity Grade Notes Research
Molecular Formula C18H15N3O2
Quantity 5 mg
Solubility Information DMSO : 50 mg/mL (163.76 mM; Need ultrasonic)
SMILES C1(NC2=CC=C(OCCO3)C3=C2)=NNC4=C1CC5=CC=CC=C45
Molecular Weight (g/mol) 305.33
Formula Weight 305.33
Percent Purity 99.0%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neurodegeneration
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.