Learn More
Sampling Systems™ GMP Pallet Knife
Product Code. 17982082 Shop All Sampling Systems Products
Change view
Click to view available options
Length (Metric):
190 mm
240 mm
325 mm
380 mm
440 mm
490 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
17982082 190 mm Each
17972122 240 mm Each
17922092 325 mm Each
17962122 380 mm Each
17912092 440 mm Each
17942092 490 mm Each
6 options
Product Code. 17982082
Brand: Sampling Systems™ A123100
Only null left
Add to basket
Add to basket
These stainless steel GMP pallet knives are ideal for a wide range of applications, including cleaning, scraping and mixing. Made from medical grade 420 stainless steel, they are crevice-free and have a polished finish for easy cleaning after use.
  • Strong and flexible
  • Range of different lengths
Specifications
Sterility Non-sterile
Autoclavable Autoclavable
Shape Curved
Disposable Reusable
Product Type Pallet Knife
Material Stainless Steel
Length (Metric) 190 mm