missing translation for 'onlineSavingsMsg'
Learn More
Learn More
GM-CSF Antibody (83308) [Janelia Fluor™ 635], Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB5181JF635
This item is not returnable.
View return policy
Description
GM-CSF Monoclonal antibody specifically detects GM-CSF in Rat samples. It is validated for Western Blot,Neutralization,Flow Cytometry (Intracellular Staining),Immunocytochemistry,CyTOF
Specifications
| GM-CSF | |
| Monoclonal | |
| Janelia Fluor 635 | |
| colony stimulating factor 2 (granulocyte-macrophage), Colony-stimulating factor, CSF, GM-CSF, GMCSFgranulocyte-macrophage colony-stimulating factor, granulocyte-macrophage colony stimulating factor, MGC131935, MGC138897, molgramostin, sargramostim | |
| E. coli-derived recombinant rat GM-CSF, Ala1-Lys127, Accession # P48750 | |
| RUO | |
| Primary | |
| Rat | |
| Liquid |
| Western Blot, Neutralization, Flow Cytometry (Intracellular Staining), Immunocytochemistry, CyTOF | |
| 83308 | |
| 50mM Sodium Borate | |
| Mouse | |
| 0.5 mL | |
| Biologically Active Proteins, Cytokine Research, Immunology | |
| 1437 | |
| Store at 4°C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction