Learn More
Glypican 1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Product Code. 16896414 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16896414 20 μL 20µL
16886414 150 μL 150µL
2 options
Product Code. 16896414
Brand: Proteintech 167001AP20UL
Rabbit Polyclonal Antibody
Glypican 1 is a cell surface heparan sulfate proteoglycan that is composed of a membrane-associated protein core substituted with a variable number of heparan sulfate chains. Members of the glypican-related integral membrane proteoglycan family (GRIPS) contain a core protein anchored to the cytoplasmic membrane via a glycosyl phosphatidylinositol linkage. These proteins may play a role in the control of cell division and growth regulation.
Specifications
Antigen Glypican 1
Applications Immunoprecipitation, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.33 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene GPC1
Gene Accession No. P35052, P35053, Q9QZF2
Gene Alias glypican, Glypican 1, GPC1
Gene Symbols Gpc1
Host Species Rabbit
Immunogen Glypican 1 Fusion Protein Ag10095
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14733, 2817, 58920
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG