Learn More
Glycyl diphenylborinate, 95%, Thermo Scientific™
Product Code. 15431688
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15431688 250 mg 250mg
15421688 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15431688 Supplier Thermo Scientific Alfa Aesar Supplier No. H55443.MD
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 14335-29-6
Molecular Formula C14H14BNO2
Molecular Weight (g/mol) 239.081
MDL Number MFCD05863277
InChI Key IVHLNYVPKWICDY-UHFFFAOYSA-N
Synonym 2-diphenylboryl oxy-2-oxoethylamine, glycyl diphenylborinate, glycine diphenylboryl ester, diphenylboramethyl 2-aminoacetate, 2-diphenylboryl oxy-2-oxoethanamine
PubChem CID 11840443
IUPAC Name diphenylboranyl 2-aminoacetate
SMILES B(C1=CC=CC=C1)(C2=CC=CC=C2)OC(=O)CN
Specifications
Quantity 250 mg
Formula Weight 239.08
Percent Purity 95%
Chemical Name or Material Glycyl diphenylborinate