Learn More
Glycolic Acid, Sigma-Aldrich™
Product Code. 15894141
Change view
Click to view available options
Quantity:
100 g
250 g
1 kg
Unit Size:
100g
1kg
250g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15894141 1 kg 1kg
15806380 100 g 100g
15884141 250 g 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 15894141 Supplier Sigma Aldrich Supplier No. 8041041000
 Restricted Item
Estimated Shipment: 19-11-2026
For synthesis
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 79-14-1
Specifications
Melting Point 74°C to 78°C
Boiling Point 100°C (decomposition)
Density 1.49 g/cm3 at 25°C
Quantity 1 kg
MDL Number MFCD00004312
Synonym Glycolic acid, Hydroxyacetic acid, Glycolic acid
Molecular Weight (g/mol) 76.05
Grade Ungraded
Chemical Name or Material Glycolic Acid