Learn More
Glycogen Phosphorylase BB/GPBB Antibody (E9B10) [DyLight 594], Novus Biologicals Biologicals™
Product Code. 30498891 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30498891 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30498891 Supplier Novus Biologicals Supplier No. NBP327945DL594
Rabbit Monoclonal Antibody
Glycogen Phosphorylase BB/GPBB Monoclonal antibody specifically detects Glycogen Phosphorylase BB/GPBB in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Antigen Glycogen Phosphorylase BB/GPBB
Applications ELISA, Western Blot
Classification Monoclonal
Clone E9B10
Conjugate DyLight 594
Formulation 50mM Sodium Borate
Gene Alias Brain glycogen phosphorylase, EC 2.4.1.1, Glycogen phosphorylase B, Glycogen phosphorylase brain form, Glycogen Phosphorylase Isoenzyme BB, MGC9213, Phosphorylase glycogen brain, PYGB
Host Species Rabbit
Immunogen Synthetic peptide derived from human Glycogen Phosphorylase BB/GPBB
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5834
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG1 κ