Learn More
Buffer Solution, pH 2.00, Chem-Lab
Product Code. 10456762
Click to view available options
Quantity:
1000 mL
Unit Size:
1 L
Product Code. 10456762
Brand: Chem-Lab CL03.0209.1000
Designed for pH measurement and maintenance in highly acidic samples. Buffer Solution, pH 2.00, Chem-Lab is a glycine solution, consisting of citrate, sodium chloride and hydrochloric acid, with a pH level of 2.00 at 20°C. Solution is traceable to standard reference materials (SRM) from NIST.
pH Level ± 0.02 at specified temperature:
  • pH 1.99 at 0°C
  • pH 1.99 at 10°C
  • pH 2.00 at 20°C
  • pH 2.00 at 25°C
  • pH 2.00 at 30°C
  • pH 2.00 at 40°C
  • pH 2.00 at 50°C
  • pH 2.00 at 60°C
  • pH 2.00 at 70°C
  • pH 2.00 at 80°C
  • pH 2.00 at 90°C
Specifications
CAS 77-92-9, 7647-14-5, 7647-01-0
Melting Point 0°C
Boiling Point 100°C
Color Colorless
Density 1.01g/mL
pH 2.00 ±0.02
Physical Form Liquid
Quantity 1000 mL
Solution Type pH Buffer
Formulation Citrate-NaCl-HCl
Odor Odorless
Traceability to NIST Yes
Solubility Information Soluble in water