Learn More
Glycine benzyl ester p-toluenesulfonate salt, 99%
Description
Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Glycine benzyl ester p-toluenesulfonate salt is a potent crosslinking inhibitor.
Solubility
Soluble in methanol. (0.1 g/mL)
Notes
Store in a cool dry place. Incompatible with oxidizing agents. Keep container tightly closed.
Specifications
Specifications
| Melting Point | 132°C to 134°C |
| Linear Formula | NH2CH2COOCH2C6H5·CH3C6H4SO3H |
| Quantity | 25 g |
| Beilstein | 3600116 |
| Solubility Information | Soluble in methanol. (0.1g/mL) |
| Formula Weight | 337.39 |
| Percent Purity | 99% |
| Chemical Name or Material | Glycine benzyl ester p-toluenesulfonate salt |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.