Learn More
Thermo Scientific Chemicals Glycine, 99%, ACS reagent
Product Code. p-4440198 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
50 g
250 g
Packaging:
Plastic Bottle
Unit Size:
250g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
12317153 50 g Plastic Bottle 50g
12327153 250 g Plastic Bottle 250g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 12317153 Supplier Thermo Scientific Chemicals Supplier No. 385900500
Request Bulk or Custom Format
CAS: 56-40-6 | C2H5NO2 | 75.07 g/mol
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
Synonym glycine, aminoacetic acid, glycocoll, aminoethanoic acid, glycolixir, glycosthene, aciport, glicoamin, padil, hampshire glycine
PubChem CID 750
ChEBI CHEBI:15428
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O
Specifications
Assay Percent Range 99%
Packaging Plastic Bottle
Linear Formula H2NCH2CO2H
Density 1.5950g/mL
Quantity 50 g
Formula Weight 75.07
Grade ACS Reagent
Chemical Name or Material Glycine