missing translation for 'onlineSavingsMsg'
Learn More

Glycine, MP Biomedicals™

Product Code. 11488272 Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 g
250 g
1 kg
3 kg
Unit Size:
1kg
250g
3kg
50g
This item is not returnable. View return policy

Product Code. 11488272

Brand: MP Biomedicals 0219404992

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Glycine, ACS Reagent Grade, White Crystalline Powder

  • It is widely used as a buffer for a variety of immunological applications.
  • It is also used as an eluent in column chromatography.
  • It has been reported for use in the enzymatic determination of lactate using lactic dehydrogenase.
  • Commonly used as a component in Tris-glycine and Tris-glycine-SDS running buffers for polyacrylamide gel electrophoresis, a component of Towbin's transfer buffer for Western blots , a buffer substance in cryoenzymology, in osmotic pressure maintenance in isoelectric focusing of erythrocytes, salting-in effect in protein chemistry, and as a buffer component in the coupled phosphatase-kinase reaction for end labelling of restriction fragments.
  • Glycine is a non-essential amino acid. Only amino acid with no asymmetric carbon.
  • It is a major inhibitory neurotransmitter.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 56-40-6
Molecular Formula C2H5NO2
Molecular Weight (g/mol) 75.07
MDL Number MFCD00008131
InChI Key DHMQDGOQFOQNFH-UHFFFAOYSA-N
IUPAC Name 2-aminoacetic acid
SMILES NCC(O)=O

Specifications

Melting Point 240°C (decomposition)(literature)
Color White
Synonym Aminoacetic acid, Glycolixir, Hampshireglycine, Aminoethanoic acid
Solubility Information Soluble in water (250mg/mL at 25°C; 672mg/mL at 100°C); slightly soluble in alcohol and ethanol(95%); almost insoluble in ether.
Density 1.16 g/mL (Lit.)
Quantity 3 kg
Grade ACS Reagent
Physical Form Crystalline Powder
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.