missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glyceryl monooleate, 36.9%, MP Biomedicals™
Shop All MP Biomedicals Products
Click to view available options
Quantity:
50 mg
1 g
Unit Size:
1g
50mg
Specifications
Specifications
| Assay Percent Range | 37% |
| Quantity | 50 mg |
| Formula Weight | 356.5 |
| Physical Form | Liquid |
| Chemical Name or Material | Glyceryl monooleate |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction