missing translation for 'onlineSavingsMsg'
Learn More

Glycerol phenylbutyrate, MedChemExpress

Product Code. p-200101300
Encompass_Preferred
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
This item is not returnable. View return policy

Product Code. 16488836

Brand: Medchem Express HYB2087

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Glycerol phenylbutyrate is a sigma-2 (σ2) receptor ligand, with a pKi of 8.02. Glycerol phenylbutyrate (GPB) is a new generation ammonia scavenger drug.

Glycerol phenylbutyrate is a sigma-2 (σ2) receptor ligand, with a pKi of 8.02. Glycerol phenylbutyrate (GPB) is a new generation ammonia scavenger drug.

Specifications

Chemical Name or Material Glycerol phenylbutyrate
CAS 611168-24-2
Color Colorless
Purity Grade Notes Research
Molecular Formula C33H38O6
Quantity 100 mg
Synonym HPN-100
Solubility Information DMSO : ≥ 150 mg/mL (282.67 mM) ∣Ethanol : 50 mg/mL (94.22 mM; Need ultrasonic)
SMILES O=C(CCCC1=CC=CC=C1)OC(COC(CCCC2=CC=CC=C2)=O)COC(CCCC3=CC=CC=C3)=O
Molecular Weight (g/mol) 530.65
Formula Weight 530.65
Percent Purity 99.18%
Grade Research
Physical Form Viscous Liquid
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.