Learn More
MP Biomedicals™ Gly-Phe-7-Amino-4-Trifluoromethylcoumarin
Product Code. 11439795 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
10mg
25mg
Unit Size:
10mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11439795 10mg 10mg
11449795 25mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11439795 Supplier MP Biomedicals™ Supplier No. 03AFC03310
  • ONLINE EXCLUSIVE PRICE
    3215.00 NOK
    3059.13 NOK / 10mg
    Save 155.87 NOK
    4% Off
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
Gly-Phe-7-Amino-4-Trifluoromethylcoumarin, White powder
  • Gly-Phe-AFC is a fluorogenic, cell-permeant peptide substrate, used in Cytotoxicity testing.
  • In vitro cytotoxicity testing has become an integral aspect of drug discovery because it is a convenient, costeffective, and predictive means of characterizing the toxic potential of new chemical entities.
  • It is also used as substrate for measuring Dipeptidyl peptidase1 (DPP1) activity.
TRUSTED_SUSTAINABILITY
Specifications
Color White
Form Powder
For Use With (Application) Cell Migration Analysis; Cell Signaling Analysis
Quantity 10mg
Molecular Weight (g/mol) 433.38 ; 574 (Including TFA salt)
Product Type Proteins, Enzymes & Peptides
Storage Requirements Keep cool and dry