missing translation for 'onlineSavingsMsg'
Learn More

GLX351322, MedChemExpress

Product Code. p-200100926
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. Quantity unitSize
16452778 10 mg 10mg
16445517 5 mg 5mg
16422526 25 mg 25mg
16414236 100 mg 100mg
16409476 50 mg 50mg
5 options
This item is not returnable. View return policy

Product Code. 16452778

Brand: Medchem Express HY100111

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GLX351322 is an inhibitor of NADPH oxidase 4 (Nox4), and inhibits hydrogen peroxide production from NOX4-overexpressing cells with an IC50 of 5 μM.

GLX351322 is an inhibitor of NADPH oxidase 4 (Nox4), and inhibits hydrogen peroxide production from NOX4-overexpressing cells with an IC50 of 5 μM.

Specifications

Chemical Name or Material GLX351322
CAS 835598-94-2
Color White
Purity Grade Notes Research
Molecular Formula C21H25N3O5S
Quantity 10 mg
Solubility Information DMSO : 20 mg/mL (46.35 mM; Need ultrasonic)
SMILES O=C(OCC)C1=C(SC2=C1CCC2)NC(CN3CCN(CC3)C(C4=CC=CO4)=O)=O
Molecular Weight (g/mol) 431.51
Formula Weight 431.51
Percent Purity 99.94%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.