missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glutathione S Transferase kappa 1 Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™
Shop All Bio Techne ProductsDescription
Glutathione S Transferase kappa 1 Polyclonal antibody specifically detects Glutathione S Transferase kappa 1 in Human samples. It is validated for ELISA,Immunocytochemistry/ Immunofluorescence
Specifications
Specifications
| Antigen | Glutathione S Transferase kappa 1 |
| Applications | ELISA, Immunocytochemistry/Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Alexa Fluor 647 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | EC 2.5.1.18, glutathione S-transferase k1, glutathione S-transferase kappa 1, Glutathione S-transferase subunit 13, glutathione S-transferase subunit 13 homolog, GST, GST 13-13, GST class-kappa, GST13, GST13-13, GSTK1-1, hGSTK1 |
| Host Species | Rabbit |
| Immunogen | Recombinant fusion protein containing a sequence corresponding to amino acids 1-226 of human Glutathione S Transferase kappa 1 (NP_057001.1).,, Sequence:, MGPLPRTVELFYDVLSPYSWLGFEILCRYQNIWNINLQLRPSLITGIMKDSGNKPPGLLPRKGLYMANDLKLLRHHLQIPIHFPKDFLSVMLEKGSLSAMRFLTAVNLEHPEMLEKASRELWMRVWSRNEDITEPQSILAAAEKAGMSAEQAQGLLEKIATPKVKNQLKETTEAACRYGAFGLPITVAHVDGQTHMLFGSDRMELLAHLLGEKWMGPIPPAVNARL |
| Purification Method | Affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?