missing translation for 'onlineSavingsMsg'
Learn More

Glutathione Oxidized, MP Biomedicals™

Product Code. 11443711 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11443711 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11443711 Supplier MP Biomedicals Supplier No. 02151193.5
  • ONLINE EXCLUSIVE PRICE
    729.00 NOK
    693.57 NOK / 500mg
    Save 35.43 NOK
    4% Off
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Protects against oxidative damage. Glutathione Oxidized, MP Biomedicals is the oxidised dimeric form of glutathione (GSH), a naturally occuring tripeptide that takes part in the body's detoxification mechanisms.

Acts as a hydrogen acceptor in enzymic determination of NADP and NADPH.

  • In vivo, Glutathione disulfide (GSSG) is reduced back to glutathione (GSH) via the NADPH-dependent enzyme glutathione reductase
  • The ratio of GSH to GSSG is often used to gauge the level of oxidative stress in cells, with higher concentrations of GSSG implying more oxidative stress

Specifications

CAS 27025-41-8
Melting Point 178°C (decomposition)(literature)
pH 1 to 6
Molecular Formula C20H32N6O12S2
MDL Number MFCD00150701
Quantity 500 mg
Synonym oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
Solubility Information Soluble in water (3.3%): Clear, Colorless Solution.
InChI Key YPZRWBKMTBYPTK-IOKZUGQQNA-N
Optical Rotation (c=1, water) –85 to –110°
SMILES N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O
IUPAC Name (2S)-2-amino-4-{[(1R)-2-{[(2R)-2-[(4S)-4-amino-4-carboxybutanamido]-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl]carbamoyl}butanoic acid
Molecular Weight (g/mol) 612.63
PubChem CID 65359
ChEBI CHEBI:17858
Formula Weight 612.64
Physical Form Crystalline Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.