missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Glutathione Monoclonal Antibody (D8)
Description
MA1-7620 detects Glutathione from all organism samples. MA1-7620 has been successfully used in ELISA, WB and IP procedures. The MA1-7620 immunogen is a Glutathione protein. Store at 4°C short term. For long term storage, store at -80°C.
Glutathione is a tripeptide comprised of three amino acids (cysteine, glutamic acid, and glycine) present in most mammalian tissue. Glutathione acts as an antioxidant, a free radical scavenger and a detoxifying agent. Glutathione is also important as a cofactor for the enzyme glutathione peroxidase, in the uptake of amino acids, and in the synthesis of leukotrienes. As a substrate for glutathione S-transferase, this agent reacts with a number of harmful chemical species, such as halides, epoxides and free radicals, to form harmless inactive products. In erythrocytes, these reactions prevent oxidative damage through the reduction of methemoglobin and peroxides. Glutathione is also involved in the formation and maintenance of disulfide bonds in proteins and in the transport of amino acids across cell membranes.
Specifications
Specifications
| Antigen | Glutathione |
| Applications | ELISA, Immunoprecipitation, Western Blot |
| Classification | Monoclonal |
| Clone | D8 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.01% sodium azide; pH 7.2 |
| Gene Alias | C10H17N3O6S; Glutathion; Glutathione-SH; GPX4; L-Glutathione; Reduced glutathione |
| Host Species | Mouse |
| Immunogen | Glutathione. |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?