missing translation for 'onlineSavingsMsg'
Learn More

Glutahione oxidized, For HPLC analysis, 98%, MP Biomedicals™

Product Code. 11433711 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11433711 250 mg 250mg
11453711 1 g 1g
11463711 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11433711 Supplier MP Biomedicals Supplier No. 02151193.2
  • ONLINE EXCLUSIVE PRICE
    651.00 NOK
    619.26 NOK / 250mg
    Save 31.74 NOK
    4% Off
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 27025-41-8
Molecular Formula C20H32N6O12S2
Molecular Weight (g/mol) 612.63
MDL Number MFCD00150701
InChI Key YPZRWBKMTBYPTK-IOKZUGQQNA-N
Synonym oxiglutatione, glutathione disulfide, gssg, oxidized glutathione, glutathione disulphide, glutathione-ssg, l-glutathione oxidized, oxigluthione, glutathone disulfide, glutathione oxidized
PubChem CID 65359
ChEBI CHEBI:17858
SMILES N[C@@H](CCC(=O)N[C@@H](CSSC[C@H](NC(=O)CC[C@H](N)C(O)=O)C(=O)NCC(O)=O)C(=O)NCC(O)=O)C(O)=O

Specifications

Color White
Quantity 250 mg
IUPAC Name (2S)-2-amino-4-{[(1R)-2-{[(2R)-2-[(4S)-4-amino-4-carboxybutanamido]-2-[(carboxymethyl)carbamoyl]ethyl]disulfanyl}-1-[(carboxymethyl)carbamoyl]ethyl]carbamoyl}butanoic acid
Formula Weight 612.64
Percent Purity 0.98
Physical Form Lyophilized Powder
Chemical Name or Material Glutahione oxidized, For HPLC analysis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.