Learn More
GLUT4 Mouse anti-Human, Unconjugated, Clone: 3G7C9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66846-1-IG-150UL
Description
The GLUT4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GLUT4. This antibody recognizes human, mouse, rat, pig antigen. The GLUT4 antibody has been validated for the following applications: IHC, WB, ELISA analysis.
Specifications
| GLUT4 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Mouse, Rat, Pig |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 3G7C9 | |
| Unconjugated | |
| 150 μL | |
| 6517 | |
| IgG2b |