Learn More
GLUD1 Mouse anti-Human, Unconjugated, Clone: 4G10D3, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 67026-1-IG-150UL
Description
The GLUD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GLUD1. This antibody recognizes human, mouse, rat antigen. The GLUD1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| GLUD1 | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 4G10D3 | |
| Unconjugated | |
| 150 μL | |
| 2746 | |
| IgG2b |