Learn More
GLUD1 Mouse anti-Human, Unconjugated, Clone: 4G10D3, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 67026-1-IG-20UL
 View more versions of this product
Product Code. 16849405
  • 1710.33 NOK / 20µL
Description
The GLUD1 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human GLUD1. This antibody recognizes human, mouse, rat antigen. The GLUD1 antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
Specifications
GLUD1
Monoclonal
1000 μg/mL
Mouse
Primary
Human, Rat, Mouse
Western Blot, Immunohistochemistry, Immunofluorescence, ELISA
4G10D3
Unconjugated
20 μL
2746
IgG2b