Learn More
Gluconic acid sodium salt, Sigma-Aldrich™
Specifications
Specifications
| Chemical Name or Material | Gluconic acid sodium salt |
| CAS | 527-07-1 |
| Melting Point | 170°C to 175°C |
| Quantity | 25 kg |
| Synonym | Gluconic acid sodium salt, Sodium gluconate, Gluconic acid sodium salt |
| Molecular Weight (g/mol) | 218.14 |
| Grade | Not Available |