missing translation for 'onlineSavingsMsg'
Learn More

Glucocorticoid receptor Rabbit anti-Human, Mouse, Rat, CoraLite 594, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech CL594-24050-100UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 17609956

  • 3828.53 NOK / 100µL
Estimated Shipment: 29-09-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Glucocorticoid receptor Polyclonal antibody apecifically detetects Glucocorticoid receptor antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence, Flow Cytometry.
Specifications

Specifications

Glucocorticoid receptor
Polyclonal
CoraLite 594
BC015610
Rabbit
Antigen affinity purification
Primary
Human, Mouse, Rat
Liquid
Immunofluorescence, Flow Cytometry
1000 μg/mL
PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3.
NR3C1
Glucocorticoid receptor fusion protein Ag21146
100 μL
2908
Store at -20°C. Avoid exposure to light.
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers