Learn More
Glucocorticoid receptor Mouse anti-Human, Unconjugated, Clone: 1D9A9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66904-1-IG-20UL
Description
The Glucocorticoid receptor antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human Glucocorticoid receptor. This antibody recognizes human, mouse, rat antigen. The Glucocorticoid receptor antibody has been validated for the following applications: IF, IHC, WB, ELISA analysis.
Specifications
| Glucocorticoid receptor | |
| Monoclonal | |
| 1000 μg/mL | |
| Mouse | |
| Primary | |
| Human, Rat, Mouse |
| Western Blot, Immunohistochemistry, Immunofluorescence, ELISA | |
| 1D9A9 | |
| Unconjugated | |
| 20 μL | |
| 2908 | |
| IgG1 |