Learn More
GLPG0634 analog, MedChemExpress
Product Code. 16448567
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16448567 5 mg 5mg
16435938 10 mg 10mg
16432087 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16448567 Supplier Medchem Express Supplier No. HY13961
MedChemExpress GLPG0634 analog (Compoun 176) is a broad spectrum JAK inhibitor with IC50 values of <100 nM against JAK1, JAK2 and JAK3.
GLPG0634 analog (Compoun 176) is a broad spectrum JAK inhibitor with IC50 values of <100 nM against JAK1, JAK2 and JAK3.
Specifications
Chemical Name or Material GLPG0634 analog
CAS 1206101-20-3
Color Khaki
Purity Grade Notes Research
Molecular Formula C23H18N6O2
Quantity 5 mg
Solubility Information DMSO : ≥ 53 mg/mL (129.13 mM)
SMILES O=C(C1CC1)NC2=NN3C(C4=CC=C(OCC5=CC=C(C#N)N=C5)C=C4)=CC=CC3=N2
Molecular Weight (g/mol) 410.43
Formula Weight 410.43
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease