missing translation for 'onlineSavingsMsg'
Learn More

GLPG0634 analog, MedChemExpress

Product Code. 16435938
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16435938 10 mg 10mg
16448567 5 mg 5mg
16432087 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16435938 Supplier Medchem Express Supplier No. HY13961

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress GLPG0634 analog (Compoun 176) is a broad spectrum JAK inhibitor with IC50 values of <100 nM against JAK1, JAK2 and JAK3.

GLPG0634 analog (Compoun 176) is a broad spectrum JAK inhibitor with IC50 values of <100 nM against JAK1, JAK2 and JAK3.

Specifications

Chemical Name or Material GLPG0634 analog
CAS 1206101-20-3
Color Khaki
Purity Grade Notes Research
Molecular Formula C23H18N6O2
Quantity 10 mg
Solubility Information DMSO : ≥ 53 mg/mL (129.13 mM)
SMILES O=C(C1CC1)NC2=NN3C(C4=CC=C(OCC5=CC=C(C#N)N=C5)C=C4)=CC=CC3=N2
Molecular Weight (g/mol) 410.43
Formula Weight 410.43
Percent Purity 98.58%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.