Learn More
GLP1 Antibody (2622G) [Biotin], Novus Biologicals™
Product Code. 30679847 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Product Code. Quantity unitSize
30679847 0.1 mL 0.10mL
1 options
Product Code. 30679847
Brand: Novus Biologicals FAB12490B
Rabbit Monoclonal Antibody
GLP1 Monoclonal antibody specifically detects GLP1 in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY
Specifications
Antigen GLP1
Applications ELISA
Classification Monoclonal
Clone 2622G
Conjugate Biotin
Formulation PBS
Gene Alias glicentin-related polypeptide, GLP1, glucagon-like peptide 1, GRPP
Host Species Rabbit
Immunogen Human GLP-1 synthetic peptide, Accession # P01275
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Signal Transduction, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 2641
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG