Learn More
Gliotoxin, TRC
Product Code. 30006061
Change view
Click to view available options
Quantity:
1 mg
5 mg
10 mg
Unit Size:
10mg
1mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30006061 5 mg 5mg
30011786 1 mg 1mg
30008500 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30006061 Supplier Toronto Research Chemicals Supplier No. TRCG4102005MG
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 67-99-2
Molecular Formula C13 H14 N2 O4 S2
Molecular Weight (g/mol) 326.39
Synonym 10H-3,10a-Epidithiopyrazino[1,2-a]indole-1,4-dione, 2,3,5a,6-tetrahydro-6-hydroxy-3-(hydroxymethyl)-2-methyl-, [3R-(3α,5aβ,6β,10aα)]- (8CI), Gliotoxin (6CI,7CI), (3R,5aS,6S,10aR)-2,3,5a,6-Tetrahydro-6-hydroxy-3-(hydroxymethyl)-2-methyl-10H-3,10a-epidithiopyrazino[1,2-a]indole-1,4-dione, Aspergillin, S. N. 12870
SMILES CN1C(=O)[C@]23CC4=CC=C[C@H](O)[C@H]4N2C(=O)[C@@]1(CO)SS3
Specifications
Quantity 5 mg
InChI Formula InChI=1S/C13H14N2O4S2/c1-14-10(18)12-5-7-3-2-4-8(17)9(7)15(12)11(19)13(14,6-16)21-20-12/h2-4,8-9,16-17H,5-6H2,1H3/t8-,9-,12+,13+/m0/s1
Formula Weight 326.0395
Percent Purity >95%
Chemical Name or Material Gliotoxin
Grade HPLC