Learn More
Glimepiride-d5, TRC
Product Code. 30404679
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
10mg
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30404679 1 mg 1mg
30389479 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30404679 Supplier Toronto Research Chemicals Supplier No. TRCG4101521MG
 Restricted Item
Estimated Shipment: 10-12-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material Glimepiride-d5
CAS 1028809-90-6
Purity Grade Notes HPLC
Molecular Formula C24H29D5N4O5S
Quantity 1 mg
Synonym 3-Ethyl-2,5-dihydro-4-methyl-N-[2-[4-[[[[trans-4-methylcyclohexyl)amino]carbonyl]amino]sulfonyl]phenyl]ethyl]-2-oxo-1H-pyrrole-1-carboxyamide-d5, Amaryl-d5
InChI Formula InChI=1S/C24H34N4O5S/c1-4-21-17(3)15-28(22(21)29)24(31)25-14-13-18-7-11-20(12-8-18)34(32,33)27-23(30)26-19-9-5-16(2)6-10-19/h7-8,11-12,16,19H,4-6,9-10,13-15H2,1-3H3,(H,25,31)(H2,26,27,30)/i1D3,4D2
SMILES O=S(C(C=C1)=CC=C1CCNC(N2C(C(C([2H])([2H])C([2H])([2H])[2H])=C(C)C2)=O)=O)(NC(N[C@H]3CC[C@H](C)CC3)=O)=O
Molecular Weight (g/mol) 495.65
Formula Weight 495.2564
Percent Purity >95