Learn More
Glecaprevir, MedChemExpress
Click to view available options
Quantity:
2 mg
5 mg
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
2mg
50mg
5mg
Description
Glecaprevir is a novel HCV NS3/4A protease inhibitor, with IC50 values ranging from 3.5 to 11.3 nM. Glecaprevir is also a SARS-CoV 3CLpro inhibitor with an IC50 of 4.09 μM.
Specifications
Specifications
| Chemical Name or Material | Glecaprevir |
| CAS | 1365970-03-1 |
| Color | White |
| Purity Grade Notes | Research |
| Molecular Formula | C38H46F4N6O9S |
| Quantity | 25 mg |
| Synonym | ABT-493 |
| Solubility Information | DMSO : ≥ 83.3 mg/mL (99.30 mM) |
| SMILES | O=C([C@H]1N(C([C@@H](NC(O[C@@]2([H])[C@@]3([H])CCC2)=O)C(C)(C)C)=O)C[C@@](OC4=NC5=CC=CC=C5N=C4C(F)(/C=C/CO3)F)([H])C1)N[C@]6([C@H](C(F)F)C6)C(NS(=O)(C7(CC7)C)=O)=O |
| Molecular Weight (g/mol) | 838.87 |
| Show More |
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.