Description
Detect and identify gas leaks quickly with the Thermo Scientific™ GLD Pro Gas Leak Detector. Specifically designed for use with GC systems, it detects minute leaks of a wide range of laboratory gases. This hand-held, easy-to-use detector features both visual and audible alarms and utlilizes an LED light display to indicate the intensity of the leak. The small probe detector is designed to reach difficult and confined locations.
- Pushbutton on/off switch
- Pushbutton zero function
- Automatic shutoff (five minutes)
- Rechargable battery (up to six hours operation)
- Durable storage case
- Probe holder
- One-year warranty
Specifications
Specifications
| For Use With (Application) | GC |
| For Use With (Equipment) | Gas Chromatography Systems |
| Description | GLD Pro Gas Leak Detector; Auto shutoff; 6-hr rechargable battery |
| Type | Gas Leak Detector |