missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glassy carbon splinter powder, 630 to 1000μm, type 2, Thermo Scientific™
Click to view available options
Quantity:
50 g
250 g
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
Packaging | Plastic bottle |
Quantity | 50 g |
UN Number | UN1325 |
Chemical Name or Material | Glassy carbon splinter powder |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'