missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Glassy carbon splinter powder, 20-50 micron, type 1, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
250 g
Description
Glassy carbon splinter powder is used as such in purifying metals, in electrodes, electrical devices & steel.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Spécification
Packaging | Glass bottle |
Quantity | 10 g |
UN Number | UN1325 |
Solubility Information | Insoluble in water. |
Chemical Name or Material | Glassy carbon splinter powder |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'