Learn More
Thermo Scientific Chemicals Glassy Carbon Conical Crucible;Vol (ml), 30;Top Dia (mm), 44;Bottom Dia (mm), 22;Ht (mm), 45
Product Code. 15463675
Change view
Click to view available options
Capacity (Metric):
10 mL
20 mL
30 mL
60 mL
100 mL
Diameter (Metric) Outer:
31mm
35mm
44mm
52mm
70mm
Height (Metric):
27mm
38mm
45mm
53mm
56mm
Unit Size:
Each
Product Code. Capacity (Metric) Diameter (Metric) Outer Height (Metric) unitSize
15463675 30 mL 44mm 45mm Each
15443645 100 mL 70mm 53mm Each
15443675 10 mL 31mm 27mm Each
15453675 20 mL 35mm 38mm Each
15473675 60 mL 52mm 56mm Each
5 options
Product Code. 15463675
Brand: Thermo Scientific Chemicals 039005.KT
Only null left
Add to basket
Glassy carbon offers high purity, corrosion resistance, thermal stability and a structure impermeable to both gases and liquids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Subjected to a high temperature heat treatment which imparts special material properties including significantly improved corrosion resistance and strength.
Glassy carbon offers:
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
TRUSTED_SUSTAINABILITY
Specifications
Description Conical
Material Glassy Carbon
Capacity (Metric) 30 mL
Diameter (Metric) Outer 44mm
Height (Metric) 45mm
Product Type Glassy Carbon Crucible
Diameter (Metric) Base 22mm