Learn More
Heidolph™ Glassware Set - Diagonal Condenser (Standard Distillations)
Product Code. 10544362 Shop All Heidolph Instruments Products
Change view
Click to view available options
Material (Coating):
Transparent plastic
Uncoated
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material (Coating) Unit Size
10544362 Uncoated Each
10351252 Transparent plastic Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 10544362 Supplier Heidolph™ Supplier No. 5130010000
Estimated Shipment: 08-01-2027
Diagonal condenser for all standard distillations is the option with the most affordable price
  • Glassware sets with a transparent plastic coating are suitable for a temperature range up to max. 80™C
  • Glassware set includes (1) 1000 mL evaporating flask and (1) 1000 mL receiving flask
  • Evaporating flasks and vapor tubes come with a standard NS 29/32 joint size but are also available with NS 24/29
  • Ventilation cap without ground joints for easy handling and grease-free operation included
  • A venting valve with refill function is optionally available
  • Not suitable for the automated distillation program Dynamic AUTOaccurate on Hei-VAP Expert/Ultimate Control models
  • Also available upon request: glassware sets for Soxhlet extraction and descending condenser system
TRUSTED_SUSTAINABILITY
Specifications
Material Glass
Type Glassware Set
For Use With (Equipment) Rotary Evaporators
Includes Glassware set G1 - Diagonal condenser (all standard distillations), 1000 mL evaporating flask and 1000 mL receiving flask
Material (Coating) Uncoated
Volume (Metric) 1000 mL