Learn More
Cole-Parmer™ Glass Weighing Micro Funnels
Product Code. 11795308 Shop All Cole-Parmer Products
Change view
Click to view available options
Material:
Borosilicate Glass
Unit Size:
Pack of 6
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Material Unit Size
11795308 Borosilicate Glass Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 11795308 Supplier Cole-Parmer™ Supplier No. 3470000
Estimated Shipment: 25-11-2026
Ensure complete sample transfer to volumetric flasks with these reusable borosilicate glass weighing funnels. Cole-Parmer™ Glass Weighing Micro Funnels are flat on one side so you can set them on a balance.
Micro funnel is sized to fit microwell plates.
Specifications
Material Borosilicate Glass
For Use With (Application) Sample Transfer to Volumetric Flasks
Product Type Weighing Funnel