Learn More
Hirschmann™ Class A Volumetric Flasks with NS Ground
Product Code. 10616332
Change view
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
19/32 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
34/35 NS Standard Ground
45/40 NS Standard Ground
7/16 NS Standard Ground
Unit Size:
Each
Pack of 2
Product Code. Capacity (Metric) Standard Taper Joint unitSize
10616332 25 mL 12/21 NS Standard Ground Pack of 2
10543841 10 L 45/40 NS Standard Ground Each
10771761 1000 mL 19/32 NS Standard Ground Each
10606141 1000 mL 24/29 NS Standard Ground Each
13167949 100 mL 12/21 NS Standard Ground Pack of 2
11836993 100 mL 14/23 NS Standard Ground Pack of 2
11891583 10 mL 7/16 NS Standard Ground Pack of 2
11801593 10 mL 10/19 NS Standard Ground Pack of 2
10781761 2000 mL 29/32 NS Standard Ground Each
10534001 200 mL 14/23 NS Standard Ground Pack of 2
10780831 20 mL 10/19 NS Standard Ground Pack of 2
10315142 20 mL 12/21 NS Standard Ground Pack of 2
11846993 250 mL 14/23 NS Standard Ground Pack of 2
10618441 25 mL 10/19 NS Standard Ground Pack of 2
10380051 5 L 34/35 NS Standard Ground Each
10498090 500 mL 19/26 NS Standard Ground Pack of 2
12608675 50 mL 12/21 NS Standard Ground Pack of 2
12618675 50 mL 14/23 NS Standard Ground Pack of 2
10044240 5 mL 7/16 NS Standard Ground Pack of 2
19 options
Product Code. 10616332
Brand: Hirschmann™ 2820371
Only null left
Add to basket
Add to basket
Allows for precise, safe liquid measurement and handling in the laboratory. Hirschmann Instruments™ Class A Volumetric Flasks with NS Ground are manufactured from durable DURAN™ glass tubing.
  • With NS ground, blue graduation
  • Without stopper
  • Acc. to DIN EN ISO 1042
  • Corrosion-free and resistant to thermal shock
  • Conformity certified, with dated batch information
Specifications
Certifications/Compliance DIN EN ISO 1042
Capacity (Metric) 25 mL
Material Borosilicate Glass, Duran Glass
Height (Metric) 110 mm
Limit of Error ±0.06 mL
Standard Taper Joint 12/21 NS Standard Ground
Tolerance ±0.06 mL
Graduated Yes
Class A