Learn More
Hirschmann™ Class A Volumetric Flasks with Hollow Glass Stopper
Click to view available options
Capacity (Metric):
5 mL
10 mL
20 mL
25 mL
50 mL
100 mL
200 mL
250 mL
500 mL
1000 mL
2000 mL
5 L
10 L
Standard Taper Joint:
10/19 NS Standard Ground
12/21 NS Standard Ground
14/23 NS Standard Ground
19/26 NS Standard Ground
19/32 NS Standard Ground
24/29 NS Standard Ground
29/32 NS Standard Ground
34/35 NS Standard Ground
45/40 NS Standard Ground
7/16 NS Standard Ground
Unit Size:
Each
Pack of 2
Description
- With NS ground, hollow glass stopper
- With blue stain graduation
- Acc. to DIN EN ISO 1042
- Corrosion-free and resistant to thermal shock
- Conformity certified, with dated batch identification
Specifications
Specifications
| Certifications/Compliance | DIN EN ISO 1042 |
| Closure Material | Glass |
| Capacity (Metric) | 10 L |
| Material | Borosilicate Glass, Duran Glass |
| Height (Metric) | 570 mm |
| Limit of Error | ±2 mL |
| Standard Taper Joint | 45/40 NS Standard Ground |
| Tolerance | ±2 mL |
| Graduated | Yes |
| Class | A |