Learn More
Hirschmann™ Hollow Glass Stoppers
Product Code. 30090410
Change view
Click to view available options
Standard Taper Joint:
NS 10/19
NS 12/21
NS 19/26
NS 24/29
NS 29/32
NS 34/35
NS 45/40
NS 7/16
Unit Size:
Each
Pack of 10
Product Code. Standard Taper Joint unitSize
30090410 NS 10/19 Pack of 10
10014001 NS 34/35 Each
10678971 NS 45/40 Each
30090416 NS 7/16 Pack of 10
30090418 NS 12/21 Pack of 10
30090396 NS 19/26 Pack of 10
10238111 NS 24/29 Each
10524781 NS 29/32 Each
8 options
Product Code. 30090410
Brand: Hirschmann™ 8900360
Only null left
Add to basket
Add to basket
For the safe and secure closure of flasks, bottles, tubing and other laboratury apparatus. Hirschmann Instruments™ Hollow Glass Stoppers are manufactured from clear glass.
  • Acc. to DIN 12 252
  • Available to fit a range of sizes
Specifications
For Use With (Equipment) For tight sealing of standard ground vessels
Quantity 1/Ea.
Standard Taper Joint NS 10/19
Material Glass
Product Type Hollow Stopper