Learn More
Karl Hecht™ Drigalski Spatula
Click to view available options
Length (Metric):
146 mm
Unit Size:
Each
Description
- Manufactured from glass
- Can also be used as a stirrer
Specifications
Specifications
| Quantity | 1 Ea. |
| Product Type | Spatula |
| For Use With (Application) | For bacteriological work, even strikeouts, microscopy and dyeing |
| Width (Metric) Blade | 45 mm |
| Material | Glass |
| Length (Metric) | 146 mm |