Learn More
Honeywell Safety Products™ Solarpro, Supra-Dura™ Lens Glasses
Description
- Helps reduce solar glare and eye fatigue.
- Supra-Dura™ advanced anti-scratch coating allows lenses to resist scratching. Lenses resist scratching 5x longer than other coatings on the market (comparative lens life test performed using a Bayer abrasion test method and may vary between environment and application).
- 100% dielectric. For use in electrical environments.
- Sport-inspired wrap-around design provides superior peripheral vision and side protection.
- Soft, padded temples and nosepiece for a comfortable, non-slip fit.
- Ratcheting temples for easy lens angle adjustment and improved fit.
- Available in a variety of lens tints including mirrored, polarized and photochromic.
Specifications
Specifications
| Certifications/Compliance | EC Category PPE 2, Quality Assurance ISO 9001 / 2000, EC Certificate Number C9114.2B-D, EC Attestation Number C9114.2B-D, Certifications EN 166 |
| Lens Type | Supra-Dura™ Scratch-resistant |
| Lens Coating | Impact-resistant, Scratch-resistant |
| Description | Dielectric |
| Product Type | Safety glasses |
| For Use With (Application) | Building, Construction, Petrochemical and Utility industries. |
| Frame Color | Gray |
| No. per Pack | 1 Ea. |
| Lens Material | Polycarbonate |
| Frame Material | Nylon, TPR |