Learn More
Hirschmann™ Disposable Micro Capillary Pipettes, End-to-End
Product Code. 31781822
Change view
Click to view available options
Volume (Metric):
1 μL
2 μL
3 μL
4 μL
5 μL
6.66 μL
10 μL
20 μL
25 μL
40 μL
50 μL
100 μL
Unit Size:
Pack of 1000
Pack of 5000
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) unitSize
31781822 2 μL Pack of 5000
11787663 3 μL Pack of 1000
11797663 4 μL Pack of 1000
11767663 1 μL Pack of 1000
11777663 2 μL Pack of 1000
11707673 5 μL Pack of 1000
11730055 100 μL Pack of 1000
10720841 6.66 μL Pack of 1000
11717673 10 μL Pack of 1000
11772533 20 μL Pack of 1000
10582501 25 μL Pack of 1000
10480771 40 μL Pack of 1000
11797673 50 μL Pack of 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Product Code. 31781822 Supplier Hirschmann™ Supplier No. 9000202
Facilitates error-free capillary filling in the laboratory. Hirschmann Instruments™ Disposable Micro Capillary Pipettes, End-to-End are manufactured from durable DURAN™ glass.
  • End-to-end, with minicaps
  • Conformity certified
  • Available either heparinized or non-heparinized
  • Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Accuracy 0.5%
Certifications/Compliance ISO 7550, CE, IVD 98/79 EC
Material DURAN
Volume (Metric) 2 μL
Length (Metric) 30 to 32 mm
Type Minicap Capillary Pipette