Learn More
Hirschmann™ Disposable Micro Capillary Pipettes, End-to-End
Facilitates error-free capillary filling in the laboratory. Hirschmann Instruments™ Disposable Micro Capillary Pipettes, End-to-End are manufactured from durable DURAN™ glass.
Brand: Hirschmann™ 9000250
Additional Details : Weight : 0.23000kg
Description
- End-to-end, with minicaps
- Conformity certified
- Available either heparinized or non-heparinized
- Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
0.5% | |
DURAN | |
30 ±0.5 mm | |
Single-use |
ISO 7550, CE, IVD 98/79 EC | |
50 μL | |
Minicap Capillary Pipette |