Learn More
Hirschmann™ Disposable Micro Capillary Pipettes, End-to-End
Click to view available options
Volume (Metric):
1 μL
2 μL
3 μL
4 μL
5 μL
6.66 μL
10 μL
20 μL
25 μL
40 μL
50 μL
100 μL
Unit Size:
Pack of 1000
Description
- End-to-end, with minicaps
- Conformity certified
- Available either heparinized or non-heparinized
- Acc. to ISO 7550, CE acc. IVD 98/79 EC
Specifications
Specifications
| Accuracy | 0.5% |
| Certifications/Compliance | ISO 7550, CE, IVD 98/79 EC |
| Material | DURAN |
| Volume (Metric) | 10 μL |
| Length (Metric) | 30 to 32 mm |
| Type | Minicap Capillary Pipette |
| Disposable | Single-use |