Learn More
Velp Scientifica™ Glass Sand
Product Code. 15329564 Shop All Velp Products
Change view
Click to view available options
Capacity (Metric):
2 kg
Unit Size:
2kg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15329564 2 kg 2kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15329564 Supplier Velp Scientifica™ Supplier No. A00000089
 Restricted Item
Estimated Shipment: 30-12-2026
Velp Scientifica™ Glass Sand for use with HU 6 Hydrolysis Unit.
  • Sand for use wtih HU 6 Hydrolysis Unit
Specifications
For Use With (Equipment) HU6 Hydrolysis Unit
Product Type Glass Sand
Capacity (Metric) 2 kg
Material Glass
Quantity 2 Pack
Weight (Metric) 2 kg