Learn More
Gravis Glass Sample Bottles
Product Code. 17580616
Change view
Click to view available options
Volume (Metric):
12.5 mL
20 mL
25 mL
67 mL
96 mL
Unit Size:
Pack of 131
Pack of 140
Pack of 210
Pack of 86
Product Code. Volume (Metric) unitSize
17580616 20 mL Pack of 131
15114633 12.5 mL Pack of 210
15134633 67 mL Pack of 140
15124633 25 mL Pack of 210
15114643 96 mL Pack of 86
5 options
Product Code. 17580616
Brand: Gravis E00920 + Q30202S88
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Excellent for holding and storing small amounts of liquid samples. Gravis Glass Sample Bottles are manufactured from soda lime glass.
  • Available in a range of capacities
Specifications
Diameter (Metric) Outer 30 mm
Material Glass
Volume (Metric) 20 mL
No. per Pack 88
Quantity 88 Pk.
Height (Metric) 45 mm