Learn More
Gravis Glass Sample Bottles
Click to view available options
Volume (Metric):
12.5 mL
20 mL
25 mL
67 mL
96 mL
Unit Size:
Pack of 131
Pack of 140
Pack of 210
Pack of 86
Description
- Available in a range of capacities
Specifications
Specifications
| Material | Glass |
| Volume (Metric) | 67 mL |
| No. per Pack | 140 |
| Quantity | 140 Pk. |