Learn More
Hecht Karl™ Glass Rolled-Rim Vial
Product Code. 10758842
Change view
Click to view available options
Volume (Metric):
5 mL
10 mL
15 mL
30 mL
Height (Metric):
24 mm
41 mm
48 mm
72 mm
Diameter (Metric) Outer:
24 mm
25 mm
28 mm
No. per Pack:
108
130
154
Unit Size:
Pack of 108
Pack of 130
Pack of 154
Product Code. Volume (Metric) Height (Metric) Diameter (Metric) Outer No. per Pack unitSize
10758842 5 mL 24 mm 24 mm 154 Pack of 154
10339451 10 mL 41 mm 24 mm 154 Pack of 154
10419241 15 mL 48 mm 25 mm 130 Pack of 130
10247531 30 mL 72 mm 28 mm 108 Pack of 108
4 options
Product Code. 10758842
Brand: Hecht Karl™ 42783008
Only null left
Add to basket
Edge
Add to basket
Designed with a rolled rim. Karl Hecht™ Glass Vials are ideal for storing small volumes of liquids.
  • Manufactured with glass for optimum visibility
  • Snap cap included
  • Available in various sizes to fit laboratory needs
Specifications
Color Clear
Diameter (Metric) Outer 24 mm
Material Glass
Volume (Metric) 5 mL
No. per Pack 154
Closure Material Plastic
Closure Type Snap Cap
Quantity 154 Pk.
Height (Metric) 24 mm